Palmitoyl chloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A13812.22
A13812.36
A13812.22EA
2330
CZK
A13812.22
A13812.36
Palmitoyl chloride ≥98%
Palmitoyl chloride
Purity: 98%
CAS No.: 112-67-4
Molecular Formula: C16H31ClO
Formula Weight: 274.88
Flash Point: 182°(359°F)
Refractive Index: 1.4520
Melting Point: ca 9-12°
Boiling Point: 174°/0.5mm
Beilstein Reference: 972409
UN No.: 3265
MDL No.: MFCD00000742
Vzorec:
C₁₆H₃₁ClO MW: 274,87 g/mol Bod varu: 174 °C (0,5 mmHg) Bod tání: ∼ 9…12 °C Hustota: 0,906 Bod vzplanutí: 182 °C (359 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000742 CAS číslo: 112-67-4 EINECS: 203-996-7 UN: 3265 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...