Lithium acetoacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Oxobutyric acid Lithium salt, Lithium 3-Oxobutyrate, Acetoacetic acid Lithium salt
A8509-10MG
A8509-250MG
A8509-1G
A8509-5G
SIALA8509-10MGEA
1370
CZK
SIALA8509-10MG
SIALA8509-250MG
SIALA8509-1G
SIALA8509-5G
Lithium acetoacetate, Sigma-Aldrich®
Lithium acetoacetate
Vzorec:
C₄H₅LiO₃ MW: 108.02 |
MDL Number:
MFCD00013060 CAS číslo: 3483-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...