Methyl acetoacetate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Methyl 3-oxobutanoate
A13696.0B
A13696.0I
A13696.30
A13696.0BEA
1980
CZK
A13696.0B
A13696.0I
A13696.30
Methyl acetoacetate ≥99%
Methyl acetoacetate
Purity: 99%
CAS No.: 105-45-3
Molecular Formula: C5H8O3
Formula Weight: 116.12
Flash Point: 70°(158°F)
Refractive Index: 1.4185
Odour: Pleasant
Melting Point: -80°
Boiling Point: 170°
Solubility: Soluble in water
Beilstein Reference: 506727
Merck Reference: 14,6009
MDL No.: MFCD00008784
Vzorec:
C₅H₈O₃ MW: 116.12 Bod varu: 170 °C Bod tání: –80 °C Hustota: 1.076 Bod vzplanutí: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008784 CAS číslo: 105-45-3 EINECS: 203-299-8 Merck Index: 12,06090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...