Isopropyl acetoacetate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Isopropyl 3-oxobutanoate
MQ200
689092-1KG
689092-230KG
689092-25KG
SIAL689092-1KGEA
5190
CZK
SIAL689092-1KG
SIAL689092-230KG
SIAL689092-25KG
Isopropyl acetoacetate, Sigma-Aldrich®
Isopropyl acetoacetate
Vzorec:
C₇H₁₂O₃ MW: 144.17 |
MDL Number:
MFCD00043933 CAS číslo: 542-08-5 EINECS: 208-798-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...