Trichloroacetyl chloride for synthesis, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
8.08391.0250EA
3860
CZK
8.08391.9999
8.08391.0250
8.08391.2500
8.08391.0100
Trichloroacetyl chloride for synthesis, Sigma-Aldrich®
Trichloroacetyl chloride
Vzorec:
C₂Cl₄O MW: 181,83 g/mol Bod varu: 118 °C (1013 hPa) Bod tání: –57 °C Hustota: 1,62 g/cm³ (20 °C) |
MDL Number:
MFCD00000792 CAS číslo: 76-02-8 EINECS: 200-926-7 UN: 2442 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...