Tisk…

1,10-Fenantrolin monohydrát ACS

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1,10-Phenanthroline hydrate

30910.09 30910.18
30910.09EA 2350 CZK
30910.09 30910.18
1,10-Fenantrolin monohydrát ACS
1,10-Fenantrolin monohydrát

CAS No.: 5144-89-8 Molecular Formula: C12H8N2.H2O Formula Weight: 198.23 (180.21anhy) Physical Form: Crystalline Odour: Odourless Melting Point: 97-101° Beilstein Reference: 4008096 Merck Reference: 14,7214 UN No.: 2811 MDL No.: MFCD00011678

Vzorec: C₁₂H₈N₂·1H₂O
MW: 198,22 g/mol
Bod tání: 97…101 °C
Hustota: 1,259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS číslo: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,7214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR