Tisk…

(R)-(+)-1-Phenylethyl isocyanate, Supelco®

Dodavatel: Merck

Synonyms: (R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate

M-Clarity™ quality level = MQ100

77968-1ML 77968-5ML
77968-1MLEA 5750 CZK
77968-1ML 77968-5ML
(R)-(+)-1-Phenylethyl isocyanate, Supelco®
(R)-(+)-1-Phenylethyl isocyanate
Vzorec: C₉H₉NO
MW: 147,18 g/mol
Bod varu: 74…76 °C (7 hPa)
Hustota: 1,05 g/cm³ (20 °C)
Bod vzplanutí: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739387
CAS číslo: 33375-06-3
EINECS: 251-485-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b0a620dd65d75f7889c271ce61478256b704a4&vid=aa3c2beb24a08e8de4464ba43e6e2885b9d36ff9&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR