(R)-(+)-1-Phenylethyl isocyanate ≥99%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
(R)-(+)-α-Phenethyl isocyanate, (R)-(+)-1-Phenethyl isocyanate, (R)-(+)-1-phenylethylisocyanate, (R)-(+)-α-Methylbenzyl isocyanate
L10305.03
L10305.06
L10305.03EA
1450
CZK
L10305.03
L10305.06
(R)-(+)-1-Phenylethyl isocyanate ≥99%
(R)-(+)-1-Phenylethyl isocyanate
Purity: 99%
CAS No.: 33375-06-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5124
Boiling Point: 55-56°/2.5mm
Beilstein Reference: 3196930
Optical Rotation: +10.5° (neat)
UN No.: 2206
MDL No.: MFCD00063015
Vzorec:
C₉H₉NO MW: 147,18 g/mol Bod varu: 55…56 °C (2,5 mmHg) Hustota: 1,045 Bod vzplanutí: 65 °C (149 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD06739387 CAS číslo: 33375-06-3 EINECS: 251-485-2 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...