Tisk…

Methylene blue hydrate 96+%, high purity biological stain

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Methylthioninium chloride hydrate, Basic Blue 9, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate, C.I. 52015, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride xhydrate

229801000 229800010
ACRO229801000EA 2580 CZK
ACRO229801000 ACRO229800010
Methylene blue hydrate 96+%, high purity biological stain
Methylene blue hydrate
Vzorec: C₁₆H₁₈ClN₃S·xH₂O
MW: 337.87
Bod tání: 100…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00150006
CAS číslo: 122965-43-9
EINECS: 200-515-2

Objednat

Výsledky specifikačního testu

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
Loss on drying 8.0 % to 22.0 % (105°C)
Dye content 96.0 to 103.0 % (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR