Rhodium(III) sulphate tetrahydrate ≥99.99% (metals basis) Rh ≥ 35.9%, Premion®
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Dirhodium trisulphate tetrahydrate
41016.01
41016.03
41016.06
41016.01EA
8170
CZK
41016.01
41016.03
41016.06
Rhodium(III) sulphate tetrahydrate ≥99.99% (metals basis) Rh ≥ 35.9%, Premion®
Rhodium(III) sulphate tetrahydrate
Purity: 99.99%
CAS No.: 15274-78-9
Molecular Formula: O12Rh2S3.4H2O
Formula Weight: 566.05 (493.99anhy)
Physical Form: Crystalline
Appearance: Brown
Odour: Odourless
UN No.: 3261
MDL No.: MFCD00798544
Vzorec:
O₁₂Rh₂S₃·4H₂O MW: 566.06 Storage Temperature: Ambient |
MDL Number:
MFCD00798544 CAS číslo: 15274-78-9 EINECS: 234-014-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...