Tisk…
Iron(III) phosphate hydrate
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Iron phosphate hydrate, Iron (III) orthophosphate hydrate
31106.30 31106.A1 31106.A7
31106.30EA 1840 CZK
31106.30 31106.A1 31106.A7
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
CAS No.: 51833-68-2 Molecular Formula: FePO4.xH2O Formula Weight: 150.82(anhy) Physical Form: Powder Appearance: Off-white
Vzorec: FeO₄P·xH₂O
MW: 168.83
Storage Temperature: Ambient
MDL Number: MFCD00016096
CAS číslo: 51833-68-2
EINECS: 233-149-7