Cobalt(II) carbonate hydrate
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
192100010
192100500
192102500
ACRO192100010EA
21070
CZK
ACRO192100010
ACRO192100500
ACRO192102500
Cobalt(II) carbonate hydrate
Cobalt(II) carbonate hydrate
Vzorec:
CCoO₃·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149654 CAS číslo: 57454-67-8 EINECS: 208-169-4 UN: 3077 ADR: 9,III |
Výsledky specifikačního testu
Appearance | Pink to red to violet Powder |
Titration Complexometric | ≥45.5 % (Cobalt) |
Cadmium (Cd) | ≤50 ppm |
Copper (Cu) | ≤100 ppm |
Iron (Fe) | ≤100 ppm |
Insoluble matter | ≤0.05 % (in HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...