Tisk…

Cobalt(II) bromide hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Cobalt dibromide hydrate, Cobaltous bromide hydrate

M-Clarity™ quality level = MQ100

334030-100G 334030-500G
SIAL334030-100GEA 3920 CZK
SIAL334030-100G SIAL334030-500G
Cobalt(II) bromide hydrate, Sigma-Aldrich®
Cobalt(II) bromide hydrate
Vzorec: Br₂Co·H₂O
MW: 236,76 g/mol
Hustota: 2,46 g/cm³ (20 °C)
MDL Number: MFCD00150208
CAS číslo: 85017-77-2
EINECS: 232-166-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR