Dusičnan-oxid zirkoničitý hydrát ≥99.9% (metals basis)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate
43224.22
43224.14
43224.36
43224.22EA
10020
CZK
43224.22
43224.14
43224.36
Dusičnan-oxid zirkoničitý hydrát ≥99.9% (metals basis)
Dusičnan-oxid zirkoničitý hydrát
Purity: 99.9%
CAS No.: 14985-18-3
Molecular Formula: ZrO(NO3)2.xH2O
Formula Weight: 231.23(anhy)
Physical Form: Crystalline
Odour: Odourless
UN No.: 1477
MDL No.: MFCD00149896
Vzorec:
N₂O₇Zr·H₂O MW: 249,25 g/mol Hustota: 2,04 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149896 CAS číslo: 14985-18-3 EINECS: 237-529-3 UN: 1477 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...