Tisk…
Zirconyl chloride hydrate ≥99.99% (trace metals basis), Sigma-Aldrich®
Dodavatel: Merck
Nebezpečí
Synonyms: Zirconium(IV) dichloride oxide hydrate, Zirconium dichloride oxide hydrate
M-Clarity™ quality level = MQ200
205028-10G 205028-50G
SIAL205028-10GEA 3060 CZK
SIAL205028-10G SIAL205028-50G
Zirconyl chloride hydrate ≥99.99% (trace metals basis), Sigma-Aldrich®
Zirconyl chloride hydrate
Vzorec: Cl₂OZr·H₂O
MW: 196.14
MDL Number: MFCD00149897
CAS číslo: 15461-27-5
EINECS: 231-717-9
UN: 3260
ADR: 8 II
Merck Index: 14,10184