Ytterbium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Dodavatel: Thermo Scientific
|
Nebezpečí
|
11196.09
11196.18
11196.09EA
4600
CZK
11196.09
11196.18
Ytterbium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium(III) nitrate hydrate
Purity: 99.99%
Molecular Formula: Yb(NO3)3.xH2O (x≈5-6)
Formula Weight: 359.05(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: White
Merck Reference: 14,10105
UN No.: 1477
MDL No.: MFCD00149883
Vzorec:
Yb(NO₃)₃·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149883 CAS číslo: 35725-34-9 EINECS: 237-384-6 UN: 1477 ADR: 5.1,III Merck Index: 14,10105 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...