Tisk…

Praseodymium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Praseodymium (III) chloride hydrate

11237.14 11237.22
11237.14EA 2770 CZK
11237.14 11237.22
Praseodymium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Praseodymium trichloride hydrate

Purity: 99.9% CAS No.: 19423-77-9 Molecular Formula: PrCl3.xH2O Formula Weight: 247.27(anhy) Physical Form: Crystalline Aggregates Odour: Odourless Merck Reference: 14,7712 MDL No.: MFCD00149828

Vzorec: Cl₃Pr·H₂O
Bod tání: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00149828
CAS číslo: 19423-77-9
EINECS: 233-794-4
Merck Index: 14,7712

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR