(1S)-(-)-Borneol ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(-)-Borneol, L(-)-Borneol, (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol
A12684.18
A12684.0B
A12684.30
A12684.18EA
1090
CZK
A12684.18
A12684.0B
A12684.30
(1S)-(-)-Borneol ≥97%
(1S)-(-)-Borneol
Purity: 97+%
CAS No.: 464-45-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 65°(149°F)
Physical Form: Cryst solid
Melting Point: 208-211°
Boiling Point: 210°
Beilstein Reference: 3587558
Merck Reference: 14,1338
Optical Rotation: -36° (c=5 in ethanol)
UN No.: 1312
MDL No.: MFCD00003759
Vzorec:
C₁₀H₁₈O MW: 154.25 Bod varu: 210 °C Bod tání: 208…211 °C Hustota: 1.011 Bod vzplanutí: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003759 CAS číslo: 464-45-9 EINECS: 207-353-1 UN: 1312 ADR: 4.1 III Merck Index: 13,01328 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...