Dodekamolybdofosforečnan triamonný hydrát, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Triammonium 12-molybdophosphate hydrate, Phosphomolybdic acid, ammonium salt hydrate, Ammonium phosphomolybdate hydrate
M-Clarity™ quality level = MQ100
342165-250G
342165-50G
SIAL342165-250GEA
14240
CZK
SIAL342165-250G
SIAL342165-50G
Dodekamolybdofosforečnan triamonný hydrát, Sigma-Aldrich®
Dodekamolybdofosforečnan triamonný hydrát
Vzorec:
H₁₂Mo₁₂N₃O₄₀P·H₂O MW: 1894.36 |
MDL Number:
MFCD00150423 CAS číslo: 54723-94-3 EINECS: 234-714-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...