Tisk…

Dodekamolybdofosforečnan triamonný hydrát, Reagent Grade

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Triammonium 12-molybdophosphate hydrate, Phosphomolybdic acid, ammonium salt hydrate, Ammonium phosphomolybdate hydrate

17984.22 17984.36
17984.22EA 2590 CZK
17984.22 17984.36
Dodekamolybdofosforečnan triamonný hydrát, Reagent Grade
Dodekamolybdofosforečnan triamonný hydrát

CAS No.: 54723-94-3 Molecular Formula: H12Mo12N3O40P.xH2O (x≈3) Formula Weight: 1876.35(anhy) Physical Form: Powder Density: 3.15 Appearance: Yellow Melting Point: dec. MDL No.: MFCD00036312

Vzorec: H₁₂Mo₁₂N₃O₄₀P·H₂O
Bod tání: Dec.
Hustota: 3,15
Storage Temperature: Ambient
MDL Number: MFCD00150423
CAS číslo: 54723-94-3
EINECS: 234-714-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR