Tisk…
Tetraethylammonium fluoride hydrate 98%
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate
216660250 216660050
ACRO216660250EA 4160 CZK
ACRO216660250 ACRO216660050
Tetraethylammonium fluoride hydrate 98%
Tetraethylammonium fluoride hydrate
Vzorec: C₈H₂₀FN·xH₂O
MW: 167.27
Storage Temperature: Ambient
MDL Number: MFCD00149990
CAS číslo: 98330-04-2
EINECS: 211-559-7
Objednat
Výsledky specifikačního testu
Appearance White to beige Crystals or adhering crystals or crystalline solid
Infrared spectrum Conforms
Water ≤28 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)