D-(-)-N-Methylglucamine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
M9179-100G
M9179-1KG
M9179-500G
SIALM9179-100GEA
5230
CZK
SIALM9179-100G
SIALM9179-1KG
SIALM9179-500G
D-(-)-N-Methylglucamine, Sigma-Aldrich®
D-(-)-N-Methylglucamine
Vzorec:
H₃CNHCH₂(CH(OH))₄CH₂OH Bod tání: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004707 CAS číslo: 6284-40-8 EINECS: 228-506-9 Merck Index: 12,06154 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...