Tisk…

L-Cystin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2

M-Clarity™ quality level = MQ200

C7602-100G C7602-10MG C7602-1KG C7602-25G
SIALC7602-100GEA 4710 CZK
SIALC7602-100G SIALC7602-10MG SIALC7602-1KG SIALC7602-25G
L-Cystin, Sigma-Aldrich®
L-Cystin
Vzorec: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Bod varu: 428 °C (1013 hPa)
Bod tání: 261…262 °C
Hustota: 1,677 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064228
CAS číslo: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR