Tisk…

Tripotassium orthophosphate monohydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Potassium phosphate monohydrate, Potassium phosphate tribasic monohydrate, Potassium phosphate hydrate

M-Clarity™ quality level = MQ200

04249-500G 04249-1KG
SIAL04249-500GEA 1750 CZK
SIAL04249-500G SIAL04249-1KG
Tripotassium orthophosphate monohydrate, Sigma-Aldrich®
Tripotassium orthophosphate monohydrate
Vzorec: K₃O₄P·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150410
CAS číslo: 27176-10-9
EINECS: 231-907-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR