Tisk…

D-Lysine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

L8021-1G L8021-25G L8021-5G
SIALL8021-1GEA 5740 CZK
SIALL8021-1G SIALL8021-25G SIALL8021-5G
D-Lysine, Sigma-Aldrich®
D-Lysine
Vzorec: C₆H₁₄N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00008234
CAS číslo: 923-27-3
EINECS: 213-091-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR