L-Homoserine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine
M-Clarity™ quality level = MQ100
H6515-10MG
H6515-1G
H6515-250MG
H6515-5G
SIALH6515-10MGEA
2040
CZK
SIALH6515-10MG
SIALH6515-1G
SIALH6515-250MG
SIALH6515-5G
L-Homoserine, Sigma-Aldrich®
L-Homoserine
Vzorec:
C₄H₉NO₃ MW: 119,12 g/mol Bod tání: 203 °C |
MDL Number:
MFCD00063090 CAS číslo: 672-15-1 EINECS: 211-590-6 Merck Index: 13,04760 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...