Tisk…

L(+)-Serin, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

M-Clarity™ quality level = MQ200

S4311-100G S4311-1KG S4311-25G
SIALS4311-100GEA 10560 CZK
SIALS4311-100G SIALS4311-1KG SIALS4311-25G
L(+)-Serin, Sigma-Aldrich®
L(+)-Serin
Vzorec: C₃H₇NO₃
MW: 105,09 g/mol
Bod varu: 259 °C (1013 hPa)
Bod tání: 215…225 °C
Hustota: 1,6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS číslo: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR