Tisk…

L(+)-Serin

Dodavatel: PanReac AppliChem

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

A1708.0100 A1708.1000
A1708.0100EA 2050 CZK
A1708.0100 A1708.1000
L(+)-Serin
L(+)-Serin
Vzorec: C₃H₇NO₃
MW: 105,09 g/mol
Bod varu: 259 °C (1013 hPa)
Bod tání: 215…225 °C
Hustota: 1,6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS číslo: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR