Hexachloroplatinic(IV) acid hydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate
M-Clarity™ quality level = MQ100
520896-1G
520896-25G
520896-5G
SIAL520896-1GEA
7000
CZK
SIAL520896-1G
SIAL520896-25G
SIAL520896-5G
Hexachloroplatinic(IV) acid hydrate, Sigma-Aldrich®
Hexachloroplatinic(IV) acid hydrate
Vzorec:
H₂Cl₆Pt·H₂O MW: 427,83 g/mol Bod tání: 60 °C |
MDL Number:
MFCD00149909 CAS číslo: 26023-84-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...