Tisk…

Hexachloroplatinic(IV) acid hydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Hydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate hydrate, Chloroplatinic acid hydrate, Dihydrogen hexachloroplatinate (IV) hydrate, Hydrogen hexachloroplatinate(IV) hydrate

M-Clarity™ quality level = MQ200

254029-250MG 254029-1G 254029-5G
SIAL254029-250MGEA 4020 CZK
SIAL254029-250MG SIAL254029-1G SIAL254029-5G
Hexachloroplatinic(IV) acid hydrate, Sigma-Aldrich®
Hexachloroplatinic(IV) acid hydrate
Vzorec: H₂Cl₆Pt·H₂O
MW: 427,83 g/mol
Bod tání: 60 °C
MDL Number: MFCD00149909
CAS číslo: 26023-84-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR