Tisk…

Methyl (S)-(-)-lactate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

M-Clarity™ quality level = MQ100

230340-100G 230340-5G
SIAL230340-100GEA 1670 CZK
SIAL230340-100G SIAL230340-5G
Methyl (S)-(-)-lactate, Sigma-Aldrich®
Methyl (S)-(-)-lactate
Vzorec: C₄H₈O₃
MW: 104,11 g/mol
Bod varu: 144 °C (1013 hPa)
Bod tání: –44 °C
Hustota: 1,09 g/cm³ (20 °C)
MDL Number: MFCD00064265
CAS číslo: 27871-49-4
EINECS: 248-704-9
UN: 3272
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR