Ethyl lactate (synthetic) ≥98% FCC, FG, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2-Hydroxypropionic Acid Ethyl Ester, Ethyl 2-Hydroxypropionate, Lactic Acid Ethyl Ester
M-Clarity™ quality level = MQ300
W244007-1KG-K
W244007-25KG-K
W244007-5KG-K
SIALW244007-1KG-KEA
2540
CZK
SIALW244007-1KG-K
SIALW244007-25KG-K
SIALW244007-5KG-K
Ethyl lactate (synthetic) ≥98% FCC, FG, Sigma-Aldrich®
Ethyl lactate
Ethyl lactate is a major volatile flavor compound identified in Chinese Luzhou-flavor type liquor.
Vzorec:
C₅H₁₀O₃ Bod varu: 154 °C (lit.) Bod tání: –25 °C Hustota: 1,031 g/mL (20 °C) (lit.) Bod vzplanutí: 46 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065359 CAS číslo: 97-64-3 EINECS: 202-598-0 UN: 1192 ADR: 3,III Merck Index: 12,03850 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...