Tisk…

Kyselina wolframfosforečná hydrát, Reagent Grade

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Phosphotungstic acid hydrate, Wolframophosphoric acid hydrate, Dodecawolframophosphoric acid hydrate, 12-Tungstophosphate hydrate, Tungstophosphoric Acid Hydrate

11827.18 11827.30
11827.18EA 3030 CZK
11827.18 11827.30
Kyselina wolframfosforečná hydrát, Reagent Grade
Kyselina wolframfosforečná hydrát

CAS No.: 12501-23-4 Molecular Formula: H3PO4.12WO3.xH2O Formula Weight: 2880.20(anhy) Physical Form: Powder Sensitivity: Moisture and Light Sensitive Appearance: Off-white to pale yellow Melting Point: 95° UN No.: 3260 MDL No.: MFCD00011342

Vzorec: H₃[P(W₃O₁₀)₄]·nH₂O
MW: 2898,07 g/mol
Bod tání: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00011342
CAS číslo: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR