Methyl (R)-(+)-lactate 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl D-(+)-Lactate, (R)-Methyl 2-hydroxypropanoate, D-(+)-Lactic acid methyll ester, Methyl (R)-(+)-2-hydroxypropionate
414490050
414490250
ACRO414490050EA
3600
CZK
ACRO414490050
ACRO414490250
Methyl (R)-(+)-lactate 98%
Methyl (R)-(+)-lactate
Vzorec:
C₄H₈O₃ MW: 104,11 g/mol Bod varu: 144 °C (1013 hPa) Hustota: 1,09 g/cm³ Bod vzplanutí: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004517 CAS číslo: 17392-83-5 EINECS: 241-420-6 UN: 3272 ADR: 3,III |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4110 to 1.4150 (20°C, 589 nm) |
GC | ≥97.5 % |
Optical rotation | +6.5° to +10.5° (20°C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...