2',4',6'-Trihydroxyacetophenone monohydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
2-Acetylphloroglucinol monohydrate, THAP monohydrate, Phloroacetophenone monohydrate
M-Clarity™ quality level = MQ100
T64602-10G
T64602-25G
SIALT64602-10GEA
3940
CZK
SIALT64602-10G
SIALT64602-25G
2',4',6'-Trihydroxyacetophenone monohydrate, Sigma-Aldrich®
2',4',6'-Trihydroxyacetophenone monohydrate
Vzorec:
C₈H₈O₄·1H₂O MW: 186,16 g/mol Bod varu: 394 °C (1013 hPa) Bod tání: 216…218 °C |
MDL Number:
MFCD00149091 CAS číslo: 207-556-5 EINECS: 207-556-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...