Tisk…

β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ400

M8250-100G M8250-1KG M8250-250G M8250-25G
SIALM8250-100GEA 6750 CZK
SIALM8250-100G SIALM8250-1KG SIALM8250-250G SIALM8250-25G
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Vzorec: C₆H₁₃NO₄S·1H₂O
Bod tání: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS číslo: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR