Tisk…
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
M-Clarity™ quality level = MQ500
M7816-1EA M7816-5X1EA
SIALM7816-1EAEA 3460 CZK
SIALM7816-1EA SIALM7816-5X1EA
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Vzorec: C₆H₁₃NO₄S·1H₂O
MW: 213.25
Bod tání: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS číslo: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929