β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
M-Clarity™ quality level = MQ400
M2933-100G
M2933-1KG
M2933-25G
M2933-500G
SIALM2933-100GEA
6750
CZK
SIALM2933-100G
SIALM2933-1KG
SIALM2933-25G
SIALM2933-500G
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Vzorec:
C₆H₁₃NO₄S·1H₂O Bod tání: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149409 CAS číslo: 145224-94-8 EINECS: 224-632-3 Merck Index: 13,05929 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...