Tisk…

β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®

Dodavatel: Merck

69892-100G 69892-25G 69892-500G
SIAL69892-100GEA 6440 CZK
SIAL69892-100G SIAL69892-25G SIAL69892-500G
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate, Sigma-Aldrich®
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Vzorec: C₆H₁₃NO₄S·1H₂O
Bod tání: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS číslo: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR