Tisk…

β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A16104.14 A16104.22 A16104.36
A16104.14EA 978 CZK
A16104.14 A16104.22 A16104.36
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate ≥98%
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate

Purity: 98% CAS No.: 145224-94-8 Molecular Formula: C6H13NO4S.H2O Formula Weight: 213.26 (195.24anhy) Physical Form: Crystalline powder Appearance: White Melting Point: ca 308° dec. Beilstein Reference: 141319 Merck Reference: 14,5902 MDL No.: MFCD00149409


Good's buffer

Vzorec: C₆H₁₃NO₄S·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS číslo: 145224-94-8
EINECS: 224-632-3
Merck Index: 14,5902

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR