Tisk…

1-Pentanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®

Dodavatel: Merck

76955-10G 76955-50G
SIAL76955-10GEA 3210 CZK
SIAL76955-10G SIAL76955-50G
1-Pentanesulphonic acid sodium salt monohydrate, Sigma-Aldrich®
1-Pentanesulphonic acid sodium salt monohydrate
Vzorec: C₅H₁₁NaO₃S·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS číslo: 207605-40-1
EINECS: 245-208-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR