1-Pentanesulphonic acid sodium salt monohydrate, Supelco®
Dodavatel: Merck
Synonyms:
Sodium 1-pentanesulphonate monohydrate
M-Clarity™ quality level = MQ100
76952-10G-F
76952-2.5G-F
76952-50G-F
76952-10G-FEA
4940
CZK
76952-10G-F
76952-2.5G-F
76952-50G-F
1-Pentanesulphonic acid sodium salt monohydrate, Supelco®
1-Pentanesulphonic acid sodium salt monohydrate
Vzorec:
C₅H₁₁NaO₃S·1H₂O MW: 192.21 Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS číslo: 207605-40-1 EINECS: 245-208-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...