1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Sodium 1-pentanesulphonate monohydrate
423200250
423201000
ACRO423200250EA
7620
CZK
ACRO423200250
ACRO423201000
1-Pentanesulphonic acid sodium salt monohydrate 98+% for HPLC
1-Pentanesulphonic acid sodium salt monohydrate
Vzorec:
C₅H₁₁NaO₃S·1H₂O MW: 192,21 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS číslo: 207605-40-1 EINECS: 245-208-4 |
Výsledky specifikačního testu
Appearance | Colorless to white Powder or crystals or flakes |
Infrared spectrum | Conforms |
UV | at 260 nm A: ≤0.01 |
Titration after Ion exchange | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...