Tisk…

1-Pentanesulphonic acid sodium salt hydrate 99%

Dodavatel: Thermo Scientific
Varování

Synonyms: Sodium 1-pentanesulphonate hydrate, n-Amylsulphonic acid, sodium salt hydrate

270370250 270371000
ACRO270370250EA 6550 CZK
ACRO270370250 ACRO270371000
1-Pentanesulphonic acid sodium salt hydrate 99%
1-Pentanesulphonic acid sodium salt hydrate
Vzorec: C₅H₁₁NaO₃S·xH₂O
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149548
CAS číslo: 1266615-65-9
EINECS: 245-208-4

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤9.6 % (K.F.)
Solubility (10 % in water) Clear colorless
Titration after Ion exchange 98.5 to 101.5 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR