Copper(II) acetate hydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
341746-100G
341746-500G
SIAL341746-100GEA
2020
CZK
SIAL341746-100G
SIAL341746-500G
Copper(II) acetate hydrate, Sigma-Aldrich®
Copper(II) acetate hydrate
Vzorec:
C₄H₆CuO₄·H₂O MW: 199.65 Storage Temperature: Ambient |
CAS číslo:
66923-66-8 EINECS: 205-553-3 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...