Tisk…

Europium triacetate hydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetic acid europium (III) salt hydrate, Europium(III) acetate xhydrate, Europium (III) acetate hydrate

M-Clarity™ quality level = MQ100

545090-10G 545090-1G
SIAL545090-10GEA 20500 CZK
SIAL545090-10G SIAL545090-1G
Europium triacetate hydrate, Sigma-Aldrich®
Europium triacetate hydrate
Vzorec: C₆H₉EuO₆·H₂O
MW: 347,11 g/mol
MDL Number: MFCD00150115
CAS číslo: 62667-64-5
EINECS: 214-670-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR