Tetrapropylammonium hydroxide 1 mol/l (1 N) in aqueous solution, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
IPC-TPA-OH
M-Clarity™ quality level = MQ200
254533-100G
254533-25G
SIAL254533-100GEA
3360
CZK
SIAL254533-100G
SIAL254533-25G
Tetrapropylammonium hydroxide 1 mol/l (1 N) in aqueous solution, Sigma-Aldrich®
Tetrapropylammonium hydroxide
Vzorec:
C₁₂H₂₉NO Storage Temperature: Refrigerator |
MDL Number:
MFCD00009360 CAS číslo: 4499-86-9 EINECS: 224-800-6 UN: 3267 ADR: 8,II |