Tisk…

Chlorid sodný 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Saline, NaCl, Sea salt, Halite

M-Clarity™ quality level = MQ200

S6546-1L S6546-4L
SIALS6546-1LEA 3720 CZK
SIALS6546-1L SIALS6546-4L
Chlorid sodný 5 mol/l (5 N) in aqueous solution, Sigma-Aldrich®
Chlorid sodný
Vzorec: NaCl
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS číslo: 7647-14-5
EINECS: 231-598-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR