Guanidinium nitrate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Guanidine nitrate
M-Clarity™ quality level = MQ200
234249-100G
234249-1KG
SIAL234249-100GEA
885
CZK
SIAL234249-100G
SIAL234249-1KG
Guanidinium nitrate, Sigma-Aldrich®
Guanidinium nitrate
Vzorec:
CH₆N₄O₃ MW: 122,08 g/mol Bod tání: 211…214 °C Hustota: 1,432 g/cm³ (20 °C) |
MDL Number:
MFCD00013028 CAS číslo: 506-93-4 EINECS: 208-060-1 UN: 1467 ADR: 5.1,III Merck Index: 13,04578 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...