Tisk…

(±)-2-Methylenanthic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B22050.06 B22050.14 B22050.22
B22050.06EA 1160 CZK
B22050.06 B22050.14 B22050.22
(±)-2-Methylenanthic acid ≥98%
(±)-2-Methylenanthic acid

Purity: 98% CAS No.: 1188-02-9 Molecular Formula: C8H16O2 Formula Weight: 144.22 Flash Point: 110°(230°F) Refractive Index: 1.4255 Boiling Point: 139-141°/30mm MDL No.: MFCD00142944

Vzorec: C₈H₁₆O₂
Bod varu: 139…141 °C (30 mmHg)
Hustota: 0,906
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00142944
CAS číslo: 1188-02-9
EINECS: 214-704-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR